EMAIL THIS PAGE TO A FRIEND
Molecular and Cellular Biochemistry
Pharmacological dose of melatonin reduces cytosolic calcium load in response to cholecystokinin in mouse pancreatic acinar cells.
PMID 25084987