EMAIL THIS PAGE TO A FRIEND
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine
Combination of bladder cancer-specific oncolytic adenovirus gene therapy with cisplatin on bladder cancer in vitro.
PMID 25085582