EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Direct large volume injection ultra-high performance liquid chromatography-tandem mass spectrometry determination of artificial sweeteners sucralose and acesulfame in well water.
PMID 25085815