EMAIL THIS PAGE TO A FRIEND
The Journal of Infectious Diseases
Prophylaxis of experimental endocarditis with antiplatelet and antithrombin agents: a role for long-term prevention of infective endocarditis in humans?
PMID 25086177