EMAIL THIS PAGE TO A FRIEND
British journal of anaesthesia
κ-Opioid receptor mediates the antinociceptive effect of nitrous oxide in mice.
PMID 25086587
Abstract
Our previous reports demonstrated that genetic deletion of μ-opioid receptor has no influence on the anaesthetic and antinociceptive effects of nitrous oxide (N2O) in mice, and that an antagonist selective for κ-opioid receptor (KOP), but not that selective for δ-opioid receptor, suppresses the antinociceptive effect of N2O. However, it is not known whether genetic deletion of KOP affects the N2O actions. We measured the minimum alveolar concentration (MAC) of volatile anaesthetics in the absence and presence of N2O. The antinociceptive action of N2O was tested by an acetic acid-writhing test and a hot-plate test. The number of c-Fos-immunopositive cells in sections from the lumbar spinal cord was counted to test whether the descending inhibitory system participates in the pharmacological action of N2O. The hypnotic action of N2O was assessed by measuring the N2O-induced decrease in the EC50 for loss of the righting reflex (EC50-LORR) of sevoflurane. Sevoflurane MAC was not significantly reduced by N2O and its antinociceptive action was almost completely abolished in KOP-knockout (KO) mice. The N2O-induced increase in c-Fos-immunopositive cells in laminae III-IV of the lumbar spinal cord was significant in wild-type (WT), but not in KOP-KO mice. In contrast, sevoflurane EC50-LORR was similarly reduced by N2O in WT and KOP-KO mice. Our study suggests that N2O demonstrates its antinociceptive action and reduces sevoflurane MAC in mice through KOP activation, whereas its hypnotic potency is not dependent on KOP activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001046 Sevoflurane, European Pharmacopoeia (EP) Reference Standard
C4H3F7O