EMAIL THIS PAGE TO A FRIEND
Human pathology
Close correlation between CXCR4 and VEGF expression and frequent CXCR7 expression in rhabdomyosarcoma.
PMID 25086956
Abstract
CXC chemokine receptor 4 (CXCR4) expression is reportedly correlated with both vascular endothelial growth factor (VEGF) expression and poor prognosis in a variety of cancers. Its relation to CXC chemokine receptor 7 (CXCR7) is also noted in several malignancies, including rhabdomyosarcoma (RMS) cell lines. However, the correlations between these chemokine receptors and angiogenic factors have not yet been adequately investigated in RMS clinical specimens. By immunohistochemistry, we assessed CXCR4, CXCR7, CC chemokine receptor 6, CC chemokine receptor 7, VEGF expression, microvessel density, and MIB-1 labeling index in 82 formalin-fixed RMS specimens, including 34 primary alveolar RMS and 44 primary embryonal RMS (ERMS). Twenty-six frozen samples were available for investigation by quantitative reverse transcription polymerase chain reaction to detect the messenger RNA expression levels of these molecules. We also evaluated their significance with respect to clinicopathological factors and patient survival rates. Primary RMS showed high expression of CXCR7 (83.1%) regardless of the histologic subtype. High cytoplasmic CXCR4 and high VEGF expression revealed significant correlations in both ERMS and alveolar RMS (P = .0051 and P = .0003, respectively). By univariate analysis of ERMS cases, the tumors with high VEGF expression showed significantly poor prognoses (P = .0017). High VEGF expression also was the independent adverse prognostic factor for ERMS. Because CXCR4, CXCR7, and VEGF are widely expressed in RMS, the combination of these antagonists may provide a potential target for molecular therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA049718
Anti-ACKR3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2702036
Anti-CCR6 (N-terminal) antibody produced in rabbit
HPA014488
Anti-CCR6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21075 Anti-CXCR4 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
C3116
Anti-CXCR4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500383
Anti-CXCR4 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2700377
Anti-CXCR7 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700198
Anti-CXCR7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS2021
Anti-DCR2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406243
Anti-PECAM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1408616
Anti-PECAM1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
V6627 Anti-Vascular Endothelial Growth Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
V1253 Anti-Vascular Endothelial Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4700463
Monoclonal Anti-CD31 antibody produced in mouse, clone MEM-05, purified immunoglobulin, buffered aqueous solution
C6598 Monoclonal Anti-CXCR4 (Fusin) antibody produced in mouse, clone 44716, purified immunoglobulin, lyophilized powder
WH0007852M2
Monoclonal Anti-CXCR4 antibody produced in mouse, clone 1F8, ascites fluid
SAB1404560
Monoclonal Anti-CXCR4, (N-terminal) antibody produced in mouse, clone 2G9, purified immunoglobulin, buffered aqueous solution
WH0004288M1
Monoclonal Anti-MKI67 antibody produced in mouse, clone 7B8, purified immunoglobulin, buffered aqueous solution
AMAB90870
Monoclonal Anti-MKI67 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1234, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300423 Monoclonal Anti-MKI67 antibody produced in mouse, clone 4A1, ascites fluid
SAB5300425 Monoclonal Anti-MKI67 antibody produced in mouse, clone 8D5, ascites fluid
WH0005175M1
Monoclonal Anti-PECAM1 antibody produced in mouse, clone 1D2-1A5, purified immunoglobulin, buffered aqueous solution
V4758 Monoclonal Anti-Vascular Endothelial Growth Factor antibody produced in mouse, clone 26503, purified immunoglobulin, lyophilized powder
SAB1402390
Monoclonal Anti-VEGF antibody produced in mouse, clone 3F7, purified immunoglobulin, buffered aqueous solution