EMAIL THIS PAGE TO A FRIEND
Biological & Pharmaceutical Bulletin
Synergistic effects of 5-fluorouracil and gambogenic acid on A549 cells: activation of cell death caused by apoptotic and necroptotic mechanisms via the ROS-mitochondria pathway.
PMID 25087947