Description
Molecular Formula
Add to Cart
561932
Deblock trichloroacetic acid solution, 3 % (w/v) in methylene chloride
C2HCl3O2
296082
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
C4H10O
91238
Diethyl ether, analytical standard
C4H10O
09-1833
Diethyl ether, for residue analysis
C4H10O
09-1855
Diethyl ether
C4H10O
W248908
Furfural, ≥98%, FCC, FG
C5H4O2
319910
Furfural, ACS reagent, 99%
C5H4O2
185914
Furfural, 99%
C5H4O2
W248924
Furfural, natural, ≥98%, FCC, FG
C5H4O2
04623
Furfural, analytical reference material
C5H4O2
396575
Perfluoropentanoic acid, 97%
C5HF9O2
672920
TCA Deblock, 3 % (w/v) in methylene chloride
C2HCl3O2
T9159
Trichloroacetic acid, BioXtra, ≥99.0%
C2HCl3O2
T6399
Trichloroacetic acid, ACS reagent, ≥99.0%
C2HCl3O2
T4885
Trichloroacetic acid, ≥99.0% (titration)
C2HCl3O2
T8657
Trichloroacetic acid, for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
C2HCl3O2
91228
Trichloroacetic acid, BioUltra, ≥99.5% (T)
C2HCl3O2
91230
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
C2HCl3O2
27242
Trichloroacetic acid, puriss., meets analytical specification of Ph. Eur., USP 21, 99-100.5% (calc. to the dried substance)
C2HCl3O2
116114
Trichloroacetic acid, reagent grade, 98%
C2HCl3O2
T0324
Trichloroacetic acid solution, ~6.25 % (w/v), 0.38 N
C2HCl3O2
T0699
Trichloroacetic acid solution, 6.1 N
C2HCl3O2
47658-U
Trichloroacetic acid solution, certified reference material, 1000 μg/mL in methyl tert-butyl ether
C2HCl3O2
224316
Zirconyl chloride octahydrate, reagent grade, 98%
Cl2OZr · 8H2O