EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
First report of Perkinsus beihaiensis in wild clams Anomalocardia brasiliana (Bivalvia: Veneridae) in Brazil.
PMID 25088443
Abstract
This is the first report of Perkinsus sp. (Bivalvia: Veneridae) infecting wild clams of the species Anomalocardia brasiliana in Brazil. The gill lamellae and rectum of 150 specimens of A. brasiliana collected in the Timonha river estuary (Ceará, Northeastern Brazil) in March 2012 were incubated in Ray's fluid thioglycollate medium (RFTM) for detection of Perkinsus sp. In RFTM, the prevalence of Perkinsus sp. was 14.7% (22/150) and the intensity of infection ranged from very light (1-10 cells across the slide) to light (12-100 cells). The presence of Perkinsus sp. was confirmed by PCR in seven (31.8%) out of 22 RFTM-positive specimens. DNA sequencing confirmed the presence of the genus Perkinsus and the phylogenetic analysis strongly indicated Perkinsus beihaiensis as the species responsible for the infection.