EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Inhibition of Escherichia coli tryptophan indole-lyase by tryptophan homologues.
PMID 25088962
Abstract
We have designed, synthesized and evaluated homotryptophan analogues as possible mechanism-based inhibitors for Escherichia coli tryptophan indole-lyase (tryptophanase, TIL, E.C. 4.1.99.1). As a quinonoid structure is an intermediate in the reaction mechanism of TIL, we anticipated that homologation of the physiological substrate, L-Trp would provide analogues resembling the transition state for β-elimination, and potentially inhibit TIL. Our results demonstrate that L-homotryptophan (1a) is a moderate competitive inhibitor of TIL, with Ki=67 μM, whereas L-bishomotryptophan (1b) displays more potent inhibition, with Ki=4.7 μM. Pre-steady-state kinetics indicated the formation of an external aldimine and quinonoid with 1a, but only the formation of an external aldimine for 1b, suggesting differences in the inhibition mechanism. These results demonstrate that formation of a quinonoid complex is not required for strong inhibition. In addition, the Trp analogues were evaluated as inhibitors of Salmonella typhimurium Trp synthase. Our results indicate that compound 1b is at least 25-fold more selective toward TIL than Trp synthase. We report that compound 1b is comparable to the most potent inhibitor previously reported, while displaying high selectivity for TIL. Thus, 1b is a potential lead for the development of novel antibacterials.
Related Materials