EMAIL THIS PAGE TO A FRIEND
Anti-Cancer Drugs
BI 6727 and GSK461364 suppress growth and radiosensitize osteosarcoma cells, but show limited cytotoxic effects when combined with conventional treatments.
PMID 25089571