PloS one
Oxygen-carbon nanotubes as a chemotherapy sensitizer for paclitaxel in breast cancer treatment.
PMID 25089613
Abstract
To study the in vivo and in vitro effects of adding oxygen carbon nanotubes (CNTs) to chemotherapy for breast cancer. MCF-7 and SK-BR-3 breast cancer cells were co-cultured with paclitaxel and then exposed to oxygen-CNTs under hypoxic conditions. Cell proliferation, viability, and apoptosis rate were analyzed. Hypoxia-inducible factor-1 alpha (HIF-1α) expression was measured using reverse transcription-polymerase chain reaction (RT-PCR) and western blot. Nude mice were used as a human breast cancer model to explore the impact of oxygen-CNTs on the in vivo chemotherapeutic effect of paclitaxel. Oxygen-CNTs had no significant effects on the growth of breast cancer cells under normoxia and hypoxia. However, in the hypoxic environment, oxygen-CNTs significantly enhanced the inhibitory effect of paclitaxel on cell proliferation, as well as the apoptosis rate. Under hypoxia, downregulation of HIF-1α and upregulation of caspase-3, caspase-8, caspase-9, LC3 and Beclin-1 were observed when paclitaxel was combined with oxygen-CNT. Furthermore, addition of oxygen-CNTs to chemotherapy was found to significantly reduce tumor weight in the tumor-bearing mice model. Oxygen-CNTs can significantly increase the chemotherapeutic effect of paclitaxel on breast cancer cells. Oxygen-CNTs may be a potential chemosensitizer in breast cancer therapy.