EMAIL THIS PAGE TO A FRIEND
ACS chemical biology
Potent antimalarial activity of acriflavine in vitro and in vivo.
PMID 25089658
Abstract
Malaria continues to be a major health problem globally. There is an urgent need to find new antimalarials. Acriflavine (ACF) is known as an antibacterial agent and more recently as an anticancer agent. Here, we report that ACF inhibits the growth of asexual stages of both chloroquine (CQ) sensitive and resistant strains of human malarial parasite, Plasmodium falciparum in vitro at nanomolar concentration. ACF clears the malaria infection in vivo from the bloodstreams of mice infected with Plasmodium berghei. Interestingly, ACF is accumulated only in the parasitized red blood cells (RBCs) and parasite specific transporters may have role in this specific drug accumulation. We further show that ACF impairs DNA replication foci formation in the parasites and affects the enzymatic activities of apicoplast specific Gyrase protein. We thus establish ACF as a potential antimalarial amidst the widespread incidences of drug resistant Plasmodium strains.