EMAIL THIS PAGE TO A FRIEND
International Journal of Food Microbiology
Bacterial maximum non-inhibitory and minimum inhibitory concentrations of different water activity depressing solutes.
PMID 25090605