EMAIL THIS PAGE TO A FRIEND
International journal of molecular medicine
Sirt3 attenuates hydrogen peroxide-induced oxidative stress through the preservation of mitochondrial function in HT22 cells.
PMID 25090966
Abstract
Sirtuins (Sirt) are a family of phylogenetically conserved nicotinamide adenine nucleotide (NAD(+))-dependent protein deacetylases, among which Sirt3 resides primarily in the mitochondria and serves as a stress responsive deacetylase, playing a role in protecting cells from damage under stress conditions. The present study aimed to investigate the role of Sirt3 in hydrogen peroxide (H(2)O(2))-induced oxidative neuronal injury in HT22 mouse hippocampal cells. Treatment with H(2)O(2) increased the expression of Sirt3 in a dose- and time-dependent manner, and the knockdown of Sirt3 using specific small interfering RNA (siRNA) exacerbated the H(2)O(2)-induced neuronal injury. The overexpression of Sirt3 induced by lentiviral transfection significantly reduced the generation of reactive oxygen species (ROS) and lipid peroxidation following injury, whereas the activities of endogenous antioxidant enzymes were not affected. Further experiments revealed that the H(2)O(2)-induced inhibition of mitochondrial complex activity and adenosine triphosphate (ATP) synthesis, the decrease in mitochondrial Ca(2+) buffering capacity and mitochondrial swelling were all partly reversed by Sirt3. Furthermore, the overexpression of Sirt3 attenuated the release of cytochrome c, the increase in the Bax/Bcl-2 ratio, as well as caspase-9/caspase-3 activity induced by H(2)O(2), and eventually inhibited apoptotic neuronal cell death. These results suggest that Sirt3 acts as a prosurvival factor, playing an essential role in protecting HT22 cells under H(2)O(2)-induced oxidative stress, possibly by inhibiting ROS accumulation and the activation of the mitochondrial apoptotic pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU093591
MISSION® esiRNA, targeting human SIRT3