EMAIL THIS PAGE TO A FRIEND
BMC Complementary and Alternative Medicine
Neuroprotective effects of melittin on hydrogen peroxide-induced apoptotic cell death in neuroblastoma SH-SY5Y cells.
PMID 25091565