EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
3,4,5-Trihydroxycinnamic acid increases heme-oxygenase-1 (HO-1) and decreases macrophage infiltration in LPS-induced septic kidney.
PMID 25091807
Abstract
We previously demonstrated that 3,4,5-trihydorxycinnamic acid (THC), a derivative of hydroxycinnamic acids, possesses protective effect in lipopolysaccharide (LPS)-induced endotoxemia models. However, the effects of THC in LPS-induced septic kidney are still unclear. Therefore, the present study was carried out to examine the effects of THC in LPS-challenged septic kidney using mesangial cell line and Balb/c mice. THC pretreatment effectively inhibited LPS-induced macrophage infiltration and the secretion of pro-inflammatory cytokines in the kidney of LPS-challenged animals. Pretreatment of rat mesangial cells with THC significantly attenuated LPS-induced PGE2 production and COX-2 expression. THC also significantly suppressed LPS-induced expression of MCP-1 in LPS-activated septic kidney and rat mesangial cells. In addition, THC significantly attenuated LPS-induced degradation of IκB-α in LPS-induced rat mesangial cells. THC also increased the expression of heme oxygenase-1 (HO-1) in LPS-challenged septic kidney and mesangial cells. Multiple signaling pathways including p38 and AKT have been observed to be involved in the THC-induced activation of HO-1 expression. The present data clearly demonstrate that THC protects LPS-challenged septic kidney by decreasing macrophage infiltration and increasing HO-1 expression, suggesting that THC might be a valuable therapeutic agent for compromised kidney in sepsis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22527F Anti-CAPG antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA019080
Anti-CAPG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA019092
Anti-CAPG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
SAB1406413
Anti-CCL2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019163
Anti-CCL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411231
Anti-CCL2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA016903
Anti-CD46 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408602
Anti-CD46, (N-terminal) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1402908
Monoclonal Anti-CCL2 antibody produced in mouse, clone 1F10, purified immunoglobulin, buffered aqueous solution
SAB1409632
Monoclonal Anti-CCL2 antibody produced in mouse, clone 2E2, purified immunoglobulin, buffered aqueous solution
SAB1404058
Monoclonal Anti-CD46 antibody produced in mouse, clone 1B6, ascites fluid
SAB4700428 Monoclonal Anti-CD46 antibody produced in mouse, clone MEM-258, purified immunoglobulin, buffered aqueous solution
WH0004179M7
Monoclonal Anti-MCP antibody produced in mouse, clone 2E10, purified immunoglobulin, buffered aqueous solution
SAB2103318
Anti-CCL2 antibody produced in rabbit, affinity isolated antibody