EMAIL THIS PAGE TO A FRIEND
Pharmaceutical Research
Characterisation of skin barrier function using bioengineering and biophysical techniques.
PMID 25092069
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
175838
Glycine-d5, 98 atom % D
C2D5NO2
396575
Perfluoropentanoic acid, 97%
C5HF9O2