EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Corosolic acid content and SSR markers in Lagerstroemia speciosa (L.) Pers.: a comparative analysis among populations across the Southern Western Ghats of India.
PMID 25092227