EMAIL THIS PAGE TO A FRIEND
Antimicrobial Agents and Chemotherapy
An upstream truncation of the furA-katG operon confers high-level isoniazid resistance in a Mycobacterium tuberculosis clinical isolate with no known resistance-associated mutations.
PMID 25092698