EMAIL THIS PAGE TO A FRIEND
Journal of Ethnopharmacology
Differentiation of bel-7402 human hepatocarcinoma cells induced by aqueous extracts of fresh gecko (AG) and its anti-tumor activity in vivo.
PMID 25093547