EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Centrally formed acetaldehyde mediates ethanol-induced brain PKA activation.
PMID 25093700
Abstract
Centrally formed acetaldehyde has proven to be responsible for several psychopharmacological effects induced by ethanol. In addition, it has been suggested that the cAMP-PKA signaling transduction pathway plays an important role in the modulation of several ethanol-induced behaviors. Therefore, we hypothesized that acetaldehyde might be ultimately responsible for the activation of this intracellular pathway. We used three pharmacological agents that modify acetaldehyde activity (α-lipoic acid, aminotriazole, and d-penicillamine) to study the role of this metabolite on EtOH-induced PKA activation in mice. Our results show that the injection of α-lipoic acid, aminotriazole and d-penicillamine prior to acute EtOH administration effectively blocks the PKA-enhanced response to EtOH in the brain. These results strongly support the hypothesis of a selective release of acetaldehyde-dependent Ca(2+) as the mechanism involved in the neurobehavioral effects elicited by EtOH.