EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Isolation of azomycin from Pseudomonas fluorescens.
PMID 2509407
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
195650
2-Nitroimidazole, 98%
C3H3N3O2