EMAIL THIS PAGE TO A FRIEND
International journal of oncology
Enhanced apoptosis by pemetrexed and simvastatin in malignant mesothelioma and lung cancer cells by reactive oxygen species-dependent mitochondrial dysfunction and Bim induction.
PMID 25096993
Abstract
Pemetrexed is a multitarget antifolate currently used for the treatment of malignant mesothelioma and non-small cell lung cancer (NSCLC). Statins, 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitors used primarily for hyperlidpidemia, have been studied for their antiproliferative and pro-apoptotic effects. However, the effects of simvastatin on pemetrexed-induced apoptosis have not been investigated. In this study, we investigated whether combination treatment with pemetrexed and simvastatin potentiates the apoptotic activity above that is seen with either drug alone in malignant mesothelioma and NSCLC cells. We found that the combination of pemetrexed and simvastatin induced more extensive caspase-dependent apoptosis than either drug alone in malignant mesothelioma cells (MSTO-211) or NSCLC cells (A549). In addition, reactive oxygen species (ROS) generation in cells treated with both pemetrexed and simvastatin was markedly increased compared to cells treated with either pemetrexed or simvastatin alone. Combination treatment also increased the loss of mitochondrial membrane potential, increased cytosolic release of cytochrome c, and altered expression of inhibitor of apoptosis proteins (IAP) and B-cell lymphoma-2 (Bcl-2) families of apoptosis related proteins. On the other hand, pretreatment with N-acetylcysteine (NAC) prevented apoptosis and mitochondrial dysfunction by pemetrexed and simvastatin. In addition, Bim siRNA conferred protection against apoptosis induced by pemetrexed and simvastatin. These results suggest that combination of pemetrexed and simvastatin potentiates their apoptotic activity beyond that of either drug alone in malignant mesothelioma and lung cancer cells. This activity is mediated through ROS-dependent mitochondrial dysfunction and Bim induction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU074771 MISSION® esiRNA, esiRNA targeting mouse Bcl2l11 (esiRNA1)