EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
Retinal ischemia/reperfusion injury is mediated by Toll-like receptor 4 activation of NLRP3 inflammasomes.
PMID 25097240
Abstract
Retinal ischemia/reperfusion (IR) is common in eye disorders. Pattern-recognition receptors (PRRs) are reported to initiate sterile inflammatory response. The role of PRRs in retinal IR injury is currently unknown. Thus, we investigated the expression and function of membrane and cytoplasmic PRRs during retinal IR. Retinal IR was induced in adult Brown Norway rats by clipping the retinal vessels for 30 minutes. RNA and proteins were extracted during the course of reperfusion, and the expression levels of the following proteins were determined: Toll-like receptor 2 (TLR2), TLR4, myeloid differentiation factor 88 (MyD88), TNF receptor-associated factor 6 (TRAF6), nuclear factor-κB (NF-κB), nucleotide-binding oligomerization domain-like receptor with pyrin domain protein 1 (NLRP1), NLRP3, apoptosis-associated speck-like protein containing a caspase recruitment domain (ASC), caspase-1, IL-1β, and IL-18. TLR4 expression in the retina was studied using immunohistochemistry. In addition, a TLR4 inhibitor was injected into the vitreous body as a therapeutic agent. After the treatment of TLR4 inhibitors, the levels of the above factors were evaluated, the apoptosis of cells in the retina, expression of cleaved-caspase-3 (c-casp-3), death of retinal ganglion cells, and the retina electroretinography was assessed. After releasing the artery clamp, the retinal vessels were reperfused in 5 minutes. During the reperfusion, TLR4, MyD88, TRAF6, NF-κB, NLRP1, NLRP3, mature IL-1β, and IL-18 were upregulated, but not TLR2. In the IR model, TLR4 was highly expressed in ganglion cell and glia cell. Additionally, the inhibition of TLR4 significantly downregulated the activation of NLRP3, but not NLRP1, and the secretion of mature IL-1β and IL-18 also were inhibited. Moreover, the TLR4 inhibitor partially attenuated the injury of the retina, including alleviated retina apoptosis, downregulated c-casp-3 expression, rescued retinal ganglion cells death, and restored retina function. These findings suggest that TLR4-signaling activation, triggered by damage-associated molecular patterns, regulates the activation of the NLRP3 inflammasomes and is responsible for the function of the retina in retinal IR injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T9778
Tropicamide, solid
C17H20N2O2
T2580000
Tropicamide, European Pharmacopoeia (EP) Reference Standard
C17H20N2O2