EMAIL THIS PAGE TO A FRIEND
Masui. The Japanese journal of anesthesiology
[A case of prolongation of rocuronium neuromuscular blockade in a pregnant patient receiving magnesium].
PMID 25098146
Abstract
A 35-year-old pregnant female with systemic lupus erythematosus and lupus nephritis underwent emergency cesarean section at 24 weeks of gestation under general anesthesia. The patient had received magnesium sulfate with a diagnosis of pregnancy-induced hypertension since 20 weeks of gestation. Anesthesia was induced with thiopental 3.5 mg x kg(-1) and tracheal intubation was facilitated by administration of rocuronium 1.0 mg x kg(-1). No additional rocuronium was needed during operation. After operation, no twitch was noted on the ulnar nerve TOF monitor. The TOF returned to 4/4 at postoperative 11 hours and the patient was extubated uneventfully. When rocuronium is used to facilitate general endotracheal anesthesia in a patient for emergency cesarean delivery, it is important to recognize that magnesium may prolong neuromuscular block significantly.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
746452
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
MgSO4
M7506
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
MgSO4
208094
Magnesium sulfate, anhydrous, reagent grade, ≥97%
MgSO4
63136
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
MgSO4
63138
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
MgO4S · 7H2O
230391
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
MgO4S · 7H2O
RES0089M-A7
Magnesium sulfate heptahydrate, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
MgO4S · 7H2O
M1880
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%
MgO4S · 7H2O
63140
Magnesium sulfate heptahydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
MgO4S · 7H2O
M5921
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
MgO4S · 7H2O
M2773
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
MgO4S · 7H2O
63142
Magnesium sulfate heptahydrate, purum p.a., ≥99.0% (T)
MgO4S · 7H2O
13142
Magnesium sulfate heptahydrate, 99.5-100.5% (calc. to the dried substance), meets analytical specification of Ph. Eur., BP,USP, FCC
MgO4S · 7H2O
R5155
Rocuronium bromide, ≥97% (perchloric acid titration)
C32H53BrN2O4
Y0001580
Rocuronium bromide, European Pharmacopoeia (EP) Reference Standard
C32H53BrN2O4
Y0000527
Rocuronium for peak identification, European Pharmacopoeia (EP) Reference Standard
C32H53BrN2O4