EMAIL THIS PAGE TO A FRIEND
Nature communications
HGK/MAP4K4 deficiency induces TRAF2 stabilization and Th17 differentiation leading to insulin resistance.
PMID 25098764
Abstract
Proinflammatory cytokines play important roles in insulin resistance. Here we report that mice with a T-cell-specific conditional knockout of HGK (T-HGK cKO) develop systemic inflammation and insulin resistance. This condition is ameliorated by either IL-6 or IL-17 neutralization. HGK directly phosphorylates TRAF2, leading to its lysosomal degradation and subsequent inhibition of IL-6 production. IL-6-overproducing HGK-deficient T cells accumulate in adipose tissue and further differentiate into IL-6/IL-17 double-positive cells. Moreover, CCL20 neutralization or CCR6 deficiency reduces the Th17 population or insulin resistance in T-HGK cKO mice. In addition, leptin receptor deficiency in T cells inhibits Th17 differentiation and improves the insulin sensitivity in T-HGK cKO mice, which suggests that leptin cooperates with IL-6 to promote Th17 differentiation. Thus, HGK deficiency induces TRAF2/IL-6 upregulation, leading to IL-6/leptin-induced Th17 differentiation in adipose tissue and subsequent insulin resistance. These findings provide insight into the reciprocal regulation between the immune system and the metabolism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7153 Anti-Insulin Receptor Substrate 1 (IRS-1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501571
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501568
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501569
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501570
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501572
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501573
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB3500281
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300482
Anti-IRS1 (Ab-636) antibody produced in rabbit, affinity isolated antibody
SAB4300483
Anti-IRS1 (Ab-639) antibody produced in rabbit, affinity isolated antibody
SAB1406045
Anti-IRS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4503887
Anti-phospho-IRS-1 (pSer1101) antibody produced in rabbit, affinity isolated antibody
SAB4504442
Anti-phospho-IRS-1 (pSer307) antibody produced in rabbit, affinity isolated antibody
SAB4504443
Anti-phospho-IRS-1 (pSer312) antibody produced in rabbit, affinity isolated antibody
SAB4300653
Anti-phospho-IRS-1 (pSer312) antibody produced in rabbit, affinity isolated antibody
SAB4503927
Anti-phospho-IRS-1 (pSer323) antibody produced in rabbit, affinity isolated antibody
SAB4503888
Anti-phospho-IRS-1 (pSer612) antibody produced in rabbit, affinity isolated antibody
SAB4504444
Anti-phospho-IRS-1 (pSer636) antibody produced in rabbit, affinity isolated antibody
SAB4504032
Anti-phospho-IRS-1 (pSer794) antibody produced in rabbit, affinity isolated antibody
SAB4504661
Anti-phospho-IRS-1 (pTyr896) antibody produced in rabbit, affinity isolated antibody
SAB4300186
Anti-phospho-IRS1 (pSer639) antibody produced in rabbit, affinity isolated antibody
SAB4300185
Anti-phospho-IRS1 (pSer636) antibody produced in rabbit, affinity isolated antibody