EMAIL THIS PAGE TO A FRIEND
The American journal of clinical nutrition
Higher fructose intake is inversely associated with risk of nonalcoholic fatty liver disease in older Finnish adults.
PMID 25099548
Abstract
High fructose intake has been suggested to be a key factor that induces nonalcoholic fatty liver disease (NAFLD), but the evidence from large epidemiologic studies is lacking. We examined the cross-sectional association between fructose intake and NAFLD by using the Fatty Liver Index (FLI) and the NAFLD liver fat score. The Helsinki Birth Cohort Study investigated 2003 Finnish men and women born in 1943-1944 in Helsinki who participated in a clinical health examination in the years 2001-2004. Trained study nurses measured weight, height, and waist circumference, and body mass index was calculated. Laboratory staff drew fasting blood for measurements of triglycerides and γ-glutamyl-transferase. The FLI and the NAFLD liver fat score were calculated on the basis of these measurements. Habitual fructose and other dietary intake over the past year were assessed by using validated and standardized 131-item food-frequency questionnaires. Data were analyzed in a cross-sectional manner by using logistic regression modeling with statistical software. In a model adjusted for age, sex, and energy intake, participants in the highest fructose intake quartile (range: 29.2-88.0 g/d) had lower risk of NAFLD assessed by using the FLI (OR: 0.56; 95% CI: 0.42, 0.75; P-trend < 0.001) and NAFLD liver fat score (OR: 0.72; 95% CI: 0.53, 0.99; P-trend < 0.001) than that of the lowest intake quartile (range: 2.2-15.2 g/d). This association remained after adjustment for educational attainment, smoking, physical activity, and other dietary variables only for the FLI (OR: 0.68; 95% CI: 0.47, 0.84; P-trend < 0.05). Our cross-sectional results did not support the current hypothesis that high intake of fructose is associated with a higher prevalence of NAFLD as assessed by using the FLI and NAFLD liver fat score.