EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim An Der Bergstrasse, Germany)
Nickel(II) complexes of pentadentate N5 ligands as catalysts for alkane hydroxylation by using m-CPBA as oxidant: a combined experimental and computational study.
PMID 25100547