EMAIL THIS PAGE TO A FRIEND
Oxidative medicine and cellular longevity
Analysis of oxidative stress enzymes and structural and functional proteins on human aortic tissue from different aortopathies.
PMID 25101153
Abstract
The role of oxidative stress in different aortopathies is evaluated. Thirty-two tissue samples from 18 men and 14 women were divided into: 4 control (C) subjects, 11 patients with systemic arterial hypertension (SAH), 4 with variants of Marfan's syndrome (MV), 9 with Marfan's syndrome (M), 2 with Turner's syndrome, and 2 with Takayasu's arteritis (TA). Aorta fragments were homogenized. Lipoperoxidation (LPO), copper-zinc and manganese superoxide dismutase (Mn and Cu-Zn-SOD), catalase (CAT), glutathione peroxidase (GPx), glutathione S-transferase (GST), endothelial nitric oxide synthase (eNOS), nitrates and nitrites (NO3(-)/NO2(-)), and type IV collagen, and laminin were evaluated. There was an increase in Mn- and Cu-Zn-SOD activity in SAH, MV, M, and Turner's syndrome. There was also an increase in CAT activity in M and Turner' syndrome. GPx and GST activity decreased and LPO increased in all groups. eNOS was decreased in SAH, MV, and M and NO3 (-)/NO2 (-) were increased in SAH and TA. Type IV collagen was decreased in Turner's syndrome and TA. Laminin γ-1 was decreased in MV and increased in M. In conclusion, similarities and differences in oxidative stress in the different aortopathies studied including pathologies with aneurysms were found with alterations in SOD, CAT, GPx, GST, and eNOS activity that modify subendothelial basement membrane proteins.