EMAIL THIS PAGE TO A FRIEND
BioMed Research International
Prevention of peritendinous adhesions using an electrospun DegraPol polymer tube: a histological, ultrasonographic, and biomechanical study in rabbits.
PMID 25101292