PLoS ONE
Dual-color fluorescence imaging to monitor CYP3A4 and CYP3A7 expression in human hepatic carcinoma HepG2 and HepaRG cells.
PMID 25101946
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7385
Demecolcine, ≥98% (HPLC)
C21H25NO5
27645
Demecolcine, ≥98.0% (sum of enantiomers, HPLC)
C21H25NO5