EMAIL THIS PAGE TO A FRIEND
JAMA dermatology
Lymphoepithelioma-like carcinoma vs inflamed squamous cell carcinoma of the skin.
PMID 25102913
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K0253
Keratin from human epidermis, aqueous solution