EMAIL THIS PAGE TO A FRIEND
Cancer Chemotherapy and Pharmacology
Pretherapeutic uracil and dihydrouracil levels in saliva of colorectal cancer patients are associated with toxicity during adjuvant 5-fluorouracil-based chemotherapy.
PMID 25102934