EMAIL THIS PAGE TO A FRIEND
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
B-cell lymphomas with concurrent MYC and BCL2 abnormalities other than translocations behave similarly to MYC/BCL2 double-hit lymphomas.
PMID 25103070
Abstract
Large B-cell lymphomas with IGH@BCL2 and MYC rearrangement, known as double-hit lymphoma (DHL), are clinically aggressive neoplasms with a poor prognosis. Some large B-cell lymphomas have concurrent abnormalities of MYC and BCL2 other than coexistent translocations. Little is known about patients with these lymphomas designated here as atypical DHL. We studied 40 patients of atypical DHL including 21 men and 19 women, with a median age of 60 years. Nine (23%) patients had a history of B-cell non-Hodgkin lymphoma. There were 30 diffuse large B-cell lymphoma (DLBCL), 7 B-cell lymphoma, unclassifiable, with features intermediate between DLBCL and Burkitt lymphoma, and 3 DLBCL with coexistent follicular lymphoma. CD10, BCL2, and MYC were expressed in 28/39 (72%), 33/35 (94%), and 14/20 (70%) cases, respectively. Patients were treated with standard (n=14) or more aggressive chemotherapy regimens (n=17). We compared the atypical DHL group with 76 patients with DHLand 35 patients with DLBCL lacking MYC and BCL2 abnormalities. The clinicopathologic features and therapies were similar between patients with atypical and typical DHL. The overall survival of patients with atypical double-hit lymphoma was similar to that of patients with double-hit lymphoma (P=0.47) and significantly worse than that of patients with DLBCL with normal MYC and BCL2 (P=0.02). There were some minor differences. Cases of atypical double-hit lymphoma more often have DLBCL morphology (P<0.01), less frequently expressed CD10 (P<0.01), and patients less often had an elevated serum lactate dehydrogenase level (P=0.01). In aggregate, these results support expanding the category of MYC/BCL2 DHL to include large B-cell lymphomas with coexistent MYC and BCL2 abnormalities other than concurrent translocations.