EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Analysis of ethyl sulfate in raw wastewater for estimation of alcohol consumption and its correlation with drugs of abuse in the city of Barcelona.
PMID 25103281