EMAIL THIS PAGE TO A FRIEND
Cancer
Monitoring of epidermal growth factor receptor tyrosine kinase inhibitor-sensitizing and resistance mutations in the plasma DNA of patients with advanced non-small cell lung cancer during treatment with erlotinib.
PMID 25103305