EMAIL THIS PAGE TO A FRIEND
Research in Veterinary Science
Murine xenograft model demonstrates significant radio-sensitising effect of liposomal doxorubicin in a combination therapy for Feline Injection Site Sarcoma.
PMID 25104320