EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
Detection of unamplified genomic DNA by a PNA-based microstructured optical fiber (MOF) Bragg-grating optofluidic system.
PMID 25104434
Abstract
Microstructured optical fibers containing microchannels and Bragg grating inscribed were internally functionalized with a peptide nucleic acid (PNA) probe specific for a gene tract of the genetically modified Roundup Ready soy. These fibers were used as an optofluidic device for the detection of DNA by measuring the shift in the wavelength of the reflected IR light. Enhancement of optical read-out was obtained using streptavidin coated gold-nanoparticles interacting with the genomic DNA captured in the fiber channels (0%, 0.1%, 1% and 10% RR-Soy), enabling to achieve statistically significant, label-free, and amplification-free detection of target DNA in low concentrations, low percentages, and very low sample volumes. Computer simulations of the fiber optics based on the finite element method (FEM) were consistent with the formation of a layer of organic material with an average thickness of 39 nm for the highest percentage (10% RR soy) analysed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3648
(3-Aminopropyl)triethoxysilane, ≥98%
C9H23NO3Si
440140
(3-Aminopropyl)triethoxysilane, 99%
C9H23NO3Si
706493
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
C9H23NO3Si
741442
(3-Aminopropyl)triethoxysilane, ≥98.0%
C9H23NO3Si
55561
Aucubin, analytical standard
C15H22O9
00090590
Aucubin, primary pharmaceutical reference standard
C15H22O9
411000
Ethanolamine, purified by redistillation, ≥99.5%
C2H7NO
398136
Ethanolamine, ACS reagent, ≥99.0%
C2H7NO
ARK2169
Ethanolamine, PharmaGrade, USP/NF, Manufactured under appropriate GMP controls for Pharma or biopharmaceutical production
C2H7NO
E9508
Ethanolamine, ≥98%
C2H7NO
02400
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
C2H7NO
E0135
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
C2H7NO
00278
Ethanolamine, analytical standard
C2H7NO
15014
Ethanolamine, ≥99%
C2H7NO
02412
Ethanolamine solution, for peptide synthesis, ~74% (T), ~70% in methanol
C2H7NO
12804
HBTU, ≥98.0% (T)
C11H16F6N5OP
387649
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
C8H19N
496219
N,N-Diisopropylethylamine, 99.5%, biotech. grade
C8H19N
D125806
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
C8H19N
03440
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
C8H19N
03439
N-Ethyldiisopropylamine solution, for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
C8H19N
T1378
Sigma 7-9®, ≥99% (titration), crystalline
C4H11NO3
RDD008
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
C4H11NO3
T6066
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, cell culture tested, ≥99.9% (titration)
C4H11NO3
T6791
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
C4H11NO3
T4661
Trizma® base, ≥99.9% (titration), crystalline
C4H11NO3
93350
Trizma® base, puriss. p.a., ≥99.7% (T)
C4H11NO3
33742
Trizma® base, puriss. p.a., buffer substance, ≥99.5%
C4H11NO3
04577
Trizma® base, tested according to Ph.Eur.
C4H11NO3
93440
Trizma® base, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
C4H11NO3