EMAIL THIS PAGE TO A FRIEND
The Journal of Clinical Investigation
Endothelial C-type natriuretic peptide maintains vascular homeostasis.
PMID 25105365
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3227
Ala-Tyr-Pro-Gly-Lys-Phe-NH2 trifluoroacetate salt, ≥98% (HPLC), solid
C34H48N8O7 · xC2HF3O2