EMAIL THIS PAGE TO A FRIEND
Journal of Neurosurgery
Is the absolute value of O(6)-methylguanine-DNA methyltransferase gene messenger RNA a prognostic factor, and does it predict the results of treatment of glioblastoma with temozolomide?
PMID 25105699