EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis and characterization of some new 4-hydroxy-coumarin derivatives.
PMID 25105917
Abstract
Some novel coumarins were synthesized starting from 4-hydroxycoumarin and methyl bromoacetate. The structures of the newly obtained compounds were confirmed by elemental analysis, mass, IR and NMR spectra.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
157910
Methyl bromoacetate, 97%
C3H5BrO2