EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Study of the matrix effects and sample dilution influence on the LC-ESI-MS/MS analysis using four derivatization reagents.
PMID 25106031
Abstract
For liquid chromatographic analysis of amino acids involving derivatization and mass-spectrometric detection, it becomes more important to evaluate the presence of matrix effects in complex samples. This is somewhat complicated for amino acid analysis where analyte free sample matrix is often unavailable. In this work, matrix effects were investigated using post-column infusion method for 9-fluorenylmethyl chloroformate (FMOC-Cl) derivatives of β-Ala, Gly and Phe and diethyl ethoxymethylenemalonate (DEEMM) derivative of β-Ala. While for DEEMM derivatives, the main signal suppression was due to the borate buffer, in case of FMOC-Cl, other FMOC-derivatives caused signal suppression. Analysis of amino acids in tea and honey with DEEMM, FMOC-Cl, p-N,N,N-trimethylammonioanilyl N'-hydroxysuccinimidyl carbamate iodide (TAHS) and dansyl chloride (DNS) showed that amino acid concentrations found with different reagents do not agree well. Sample dilution experiments indicated that the sample matrix affected the analysis results obtained with DEEMM the least, but with FMOC-Cl, TAHS and DNS, sample dilution had an influence on the results. When sample dilution and extrapolative dilution approach were applied on the latter results, an agreement of amino acid concentrations measured with different reagents was achieved within relative standard deviation (RSD) of 22% for most cases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D94208
Diethyl ethoxymethylenemalonate, 99%
C10H16O5
05689
Diethyl ethoxymethylenemalonate, for LC-MS derivatization, LiChropur, ≥98.0% (GC)
C10H16O5
PHR1106
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14N4O2
A9256
L-Aspartic acid, reagent grade, ≥98% (HPLC)
C4H7NO4
11189
L-Aspartic acid, BioUltra, ≥99.5% (T)
C4H7NO4
A7219
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C4H7NO4
A8949
L-Aspartic acid, BioXtra, ≥99% (HPLC)
C4H7NO4
A5474
L-Aspartic acid, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C4H7NO4
PHR1104
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C4H7NO4
51572
L-Aspartic acid, certified reference material, TraceCERT®
C4H7NO4
W365601
L-Aspartic acid, ≥98%, FG
C4H7NO4
H8000
L-Histidine, ReagentPlus®, ≥99% (TLC)
C6H9N3O2
H6034
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
C6H9N3O2
H3911
L-Histidine, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C6H9N3O2
53319
L-Histidine, BioUltra, ≥99.5% (NT)
C6H9N3O2
PHR1108
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
C6H9N3O2
73767
L-Histidine, certified reference material, TraceCERT®
C6H9N3O2
P2126
L-Phenylalanine, reagent grade, ≥98%
C9H11NO2
P5482
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C9H11NO2
PHR1100
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
C9H11NO2
1530503
L-Phenylalanine, United States Pharmacopeia (USP) Reference Standard
C9H11NO2
P8740
L-Phenylalanine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C9H11NO2
40541
L-Phenylalanine, certified reference material, TraceCERT®
C9H11NO2
91331
L-Phenylalanine, analytical standard, for Nitrogen Determination According to Kjeldahl Method
C9H11NO2
W358509
L-Phenylalanine, 99%, FCC
C9H11NO2
P8865
L-Proline, PharmaGrade, Ajinomoto, EP, JP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H9NO2
1568506
L-Proline, United States Pharmacopeia (USP) Reference Standard
C5H9NO2
93693
L-Proline, certified reference material, TraceCERT®
C5H9NO2
T0254
L-Tryptophan, reagent grade, ≥98% (HPLC)
C11H12N2O2
T8941
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
C11H12N2O2