EMAIL THIS PAGE TO A FRIEND
Environmental Pollution (Barking, Essex : 1987)
Particle pollution in Rio de Janeiro, Brazil: increase and decrease of pro-inflammatory cytokines IL-6 and IL-8 in human lung cells.
PMID 25106047