EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Participation of thioredoxin in the V(V)-reduction reaction by Vanabin2.
PMID 25108063
Abstract
It is well-understood that ascidians accumulate high levels of vanadium, a reduced form of V(III), in an extremely acidic vacuole in their blood cells. Vanabins are small cysteine-rich proteins that have been identified only from vanadium-rich ascidians. A previous study revealed that Vanabin2 can act as a V(V)-reductase in the glutathione cascade. AsTrx1, a thioredoxin gene, was cloned from the vanadium-rich ascidian, Ascidia sydneiensis samea, by PCR. AsTrx1 and Vanabin2 were prepared as recombinant proteins, and V(V)-reduction by Vanabin2 was assessed by ESR and ion-exchange column chromatography. Site-directed mutagenesis was performed to examine the direct involvement of cysteine residues. Tissue expression of AsTrx1 was also examined by RT-PCR. When reduced AsTrx1 and Vanabin2 were combined, Vanabin2 adopted an SS/SH intermediate structure while V(V) was reduced to V(IV). The loss of cysteine residues in either Vanabin2 or AsTrx1 caused a significant loss of reductase activity. Vapp and Kapp values for Vanabin2-catalyzed V(V)-reduction in the thioredoxin cascade were 0.066mol-V(IV)/min/mol-Vanabin2 and 0.19mM, respectively. The Kapp value was 2.7-fold lower than that observed in the glutathione cascade. The AsTrx1 gene was expressed at a very high level in blood cells, in which Vanabins 1-4 were co-expressed. AsTrx1 may contribute to a significant part of the redox cascade for V(V)-reduction by Vanabin2 in the cytoplasm of vanadocytes, but prevails only at low V(V) concentrations. This study is the first to report the reduction of V(V) in the thioredoxin cascade.