EMAIL THIS PAGE TO A FRIEND
International Journal of Biological Macromolecules
Micro- and nano-hydroxyapatite as active reinforcement for soft biocomposites.
PMID 25109458
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
450146
Calcium hydroxide, 99.995% trace metals basis
H2CaO2