EMAIL THIS PAGE TO A FRIEND
Bioconjugate chemistry
Magnetic Prussian blue nanoparticles for targeted photothermal therapy under magnetic resonance imaging guidance.
PMID 25109612
Abstract
This paper reported a core-shell nanotheranostic agent by growing Prussian blue (PB) nanoshells of 3-6 nm around superparamagnetic Fe3O4 nanocores for targeted photothermal therapy of cancer under magnetic resonance imaging (MRI) guidance. Both in vitro and in vivo experiments proved that the Fe3O4@PB core-shell nanoparticles showed significant contrast enhancement for T2-weighted MRI with the relaxivity value of 58.9 mM(-1)·s(-1). Simultaneously, the composite nanoparticles exhibited a high photothermal effect under irradiation of a near-infrared laser due to the strong absorption of PB nanoshells, which led to more than 80% death of HeLa cells with only 0.016 mg·mL(-1) of the nanoparticles with the aid of the magnetic targeting effect. Using tumor-bearing nude mice as the model, the near-infrared laser light ablated the tumor effectively in the presence of the Fe3O4@PB nanoparticles and the tumor growth inhibition was evaluated to be 87.2%. Capabilities of MRI, magnetic targeting, and photothermal therapy were thus integrated into a single agent to allow efficient MRI-guided targeted photothermal therapy. Most importantly, both PB and Fe3O4 nanoparticles were already clinically approved drugs, so the Fe3O4@PB nanoparticles as a theranostic nanomedicine would be particularly promising for clinical applications in the human body due to the reliable biosafety.