EMAIL THIS PAGE TO A FRIEND
Neural plasticity
Behavioral improvement and regulation of molecules related to neuroplasticity in ischemic rat spinal cord treated with PEDF.
PMID 25110592
Abstract
Pigment epithelium derived factor (PEDF) exerts trophic actions to motoneurons and modulates nonneuronal restorative events, but its effects on neuroplasticity responses after spinal cord (SC) injury are unknown. Rats received a low thoracic SC photothrombotic ischemia and local injection of PEDF and were evaluated behaviorally six weeks later. PEDF actions were detailed in SC ventral horn (motor) in the levels of the lumbar central pattern generator (CPG), far from the injury site. Molecules related to neuroplasticity (MAP-2), those that are able to modulate such event, for instance, neurotrophic factors (NT-3, GDNF, BDNF, and FGF-2), chondroitin sulfate proteoglycans (CSPG), and those associated with angiogenesis and antiapoptosis (laminin and Bcl-2) and Eph (receptor)/ephrin system were evaluated at cellular or molecular levels. PEDF injection improved motor behavioral performance and increased MAP-2 levels and dendritic processes in the region of lumbar CPG. Treatment also elevated GDNF and decreased NT-3, laminin, and CSPG. Injury elevated EphA4 and ephrin-B1 levels, and PEDF treatment increased ephrin A2 and ephrins B1, B2, and B3. Eph receptors and ephrins were found in specific populations of neurons and astrocytes. PEDF treatment to SC injury triggered neuroplasticity in lumbar CPG and regulation of neurotrophic factors, extracellular matrix molecules, and ephrins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59060
2-Methylbutane, analytical standard
C5H12
SAB4500696
Anti-EFNA1 antibody produced in rabbit, affinity isolated antibody
SAB1410322
Anti-EFNA1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1401123
Anti-EFNA3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4500701
Anti-EFNA5 antibody produced in rabbit, affinity isolated antibody
SAB2501389
Anti-EFNB2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4500774
Anti-EPHA1 antibody produced in rabbit, affinity isolated antibody
SAB4500731
Anti-EPHA2 antibody produced in rabbit, affinity isolated antibody
SAB1410325
Anti-EPHA2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2701156
Anti-EPHA3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900055
Anti-EPHA4 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107644
Anti-EPHA4 antibody produced in rabbit, affinity isolated antibody
HPA031433
Anti-EPHA8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500732
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
SAB4500775
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
SAB4500776
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
HPA007698
Anti-EPHB3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA008184
Anti-EPHB3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500778
Anti-EPHB4 antibody produced in rabbit, affinity isolated antibody
E7525 Anti-Ephrin-B2 antibody produced in goat, affinity isolated antibody
SAB2101795
Anti-PHGDH (ab1) antibody produced in rabbit, affinity isolated antibody
HPA021241
Anti-PHGDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA024031
Anti-PHGDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2900054
Anti-EPHA4 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100689
Anti-EPHA5 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2105669
Anti-EFNB1 antibody produced in rabbit, affinity isolated antibody