EMAIL THIS PAGE TO A FRIEND
Journal of neuropathology and experimental neurology
Arterial wall degeneration plus hemodynamic insult cause arterial wall remodeling and nascent aneurysm formation at specific sites in dogs.
PMID 25111020
Abstract
To determine whether arterial wall degeneration, in combination with hemodynamic insult, causes cerebral artery aneurysms in a dog model, we simulated the geometry and hemodynamics of a human artery by surgical reconstruction of both common carotid arteries in 12 dogs. The dogs were then randomly assigned to one of the following groups: hemodynamic insult + elastase insult group ( n = 6), hemodynamic insult group (n = 6), or elastase control group (n = 3), in which the straight common carotid arteries were subjected to elastase alone. Angiography and hemodynamic analysis were performed immediately and at 12 weeks after surgery; the animals were then killed for histologic evaluation. The 12 surgically reconstructed distal internal carotid arteries simulated the human artery well with respect to geometric and hemodynamic measurements, with the intended aneurysm sites exposed to higher wall shear stress and velocity, lower pressure, turbulent flow, and changes in wall shear stress gradient. Nascent aneurysms developed in 4 hemodynamic insult + elastase insult group dogs at 12 weeks; blood flow analysis demonstrated decreased wall shear stress, increased pressure, and wall shear stress gradient from the neck to the dome. Arterial wall remodeling or nascent aneurysm formation in the hemodynamic insult + elastase insult group versus the other groups was indicated by internal elastic lamina/elastic fiber disruption, muscular layer thinning, increased smooth muscle cell proliferation, macrophage infiltration, and high expression of matrix metalloproteinase-2 and matrix metalloproteinase-9 in the media. These data suggest that nascent aneurysms were caused by the combination of arterial wall degeneration and hemodynamic perturbations in this distal internal carotid artery dog model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA000440
Anti-CD45 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406243
Anti-PECAM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1408616
Anti-PECAM1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW21465 Anti-PTPRC antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500903
Anti-S100A9 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA004193
Anti-S100A9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411220
Anti-S100A9 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4700463
Monoclonal Anti-CD31 antibody produced in mouse, clone MEM-05, purified immunoglobulin, buffered aqueous solution
SAB4700471 Monoclonal Anti-CD45 antibody produced in mouse, clone MEM-28, purified immunoglobulin, buffered aqueous solution
AMAB90518
Monoclonal Anti-CD45 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0159, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90519
Monoclonal Anti-CD45 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0160, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700587 Monoclonal Anti-CD45 antibody produced in rat, clone EM-05, purified immunoglobulin, buffered aqueous solution
WH0005175M1
Monoclonal Anti-PECAM1 antibody produced in mouse, clone 1D2-1A5, purified immunoglobulin, buffered aqueous solution
SAB4700649 Monoclonal Anti-PTPRC antibody produced in mouse, clone IVA103, purified immunoglobulin, buffered aqueous solution
SAB1404348
Monoclonal Anti-S100A9 antibody produced in mouse, clone 4G9, purified immunoglobulin, buffered aqueous solution
SAB1409623
Monoclonal Anti-S100A9 antibody produced in mouse, clone 1C22, purified immunoglobulin, buffered aqueous solution