EMAIL THIS PAGE TO A FRIEND
Growth factors (Chur, Switzerland)
Shear stress and VEGF enhance endothelial differentiation of human adipose-derived stem cells.
PMID 25112491
Abstract
Herein we combine chemical and mechanical stimulation to investigate the effects of vascular endothelial growth factor (VEGF) and physiological shear stress in promoting the differentiation human adipose derived stem cells (ADSCs) into endothelial cells. ADSCs were isolated and characterized; endothelial differentiation was promoted by culturing confluent cells in 50 ng/ml VEGF under physiological shear stress for up to 14 days. Afterwards, endothelial cells were seeded onto collagen or acellular aortic valve matrices and exposed to four culture conditions: shear stress + VEGF; shear stress - VEGF; static + VEGF and static - VEGF. After 7 days, phenotype was investigated. ADSCs subjected to shear stress and VEGF express a comprehensive range of specific endothelial markers (vWF, eNOS and FLT-1 after 7 days and CD31, FLk-1 and VE-cadherin after 14 days) and maintain the phenotype when seeded onto scaffolds. Our protocol proved to be an efficient source of endothelial-like cells for tissue engineering based on autologous ADSC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0550
Ala-Gln, ≥98% (HPLC)
C8H15N3O4
A8185
Ala-Gln, BioReagent, suitable for cell culture, suitable for insect cell culture
C8H15N3O4
G8541
Ala-Gln, 200 mM, solution, sterile-filtered, Biotechnology Performance Certified
C8H15N3O4
GW22756 Anti-Endoglin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
E7534
Anti-Endoglin antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA011862
Anti-ENG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700258 Monoclonal Anti-CD105 antibody produced in mouse, clone MEM-226, purified immunoglobulin, buffered aqueous solution
SAB4700262 Monoclonal Anti-CD105 antibody produced in mouse, clone MEM-229, purified immunoglobulin, buffered aqueous solution
AMAB90925
Monoclonal Anti-ENG antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1912, purified immunoglobulin, buffered aqueous glycerol solution
85886
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
C21H39N7O12 · 1.5H2SO4
S1400000
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
C21H39N7O12 · 1.5H2O4S
WH0002022M1
Monoclonal Anti-ENG antibody produced in mouse, clone 4C11, purified immunoglobulin, buffered aqueous solution
AV33068
Anti-ENG (AB1) antibody produced in rabbit, IgG fraction of antiserum